Learn More
Medchemexpress LLC Cbz-B3A 1mg | 1884710-81-9 | 1 MG
SDP
Catalog No. 5000229837
Encompass_Preferred
Cbz-B3A is a small-molecule research inhibitor of mTORC1 signaling that binds ubiquilins and inhibits phosphorylation of eIF4E-binding protein 1 (4EBP1), reducing cap-dependent translation in cell-based assays. Supplied as a solid for in vitro research.
  • Potent, selective mTORC1 signaling inhibitor.
  • Inhibits phosphorylation of eIF4E-binding protein 1 (4EBP1).
  • High purity solid powder (≈98.6%).
  • Soluble in DMSO at 20 mg/mL; warming and sonication may be required.
  • Intended for in vitro research use only.
Catalog No. 50-002-29837 Supplier Medchemexpress LLC Supplier No. HY1142671MG
Cbz-B3A is a small-molecule research inhibitor of mTORC1 signaling that binds ubiquilins and inhibits phosphorylation of eIF4E-binding protein 1 (4EBP1), reducing cap-dependent translation in cell-based assays. Supplied as a solid for in vitro research.
  • Potent, selective mTORC1 signaling inhibitor.
  • Inhibits phosphorylation of eIF4E-binding protein 1 (4EBP1).
  • High purity solid powder (≈98.6%).
  • Soluble in DMSO at 20 mg/mL; warming and sonication may be required.
  • Intended for in vitro research use only.