Learn More
Cayman Chemical Cbz-Gly-Pro-AMC, Purity: ≥98%, CAS: 68542-93-8, 10 mg
SDP
Catalog No. 502944036
Encompass_Preferred
A fluorogenic substrate for prolyl endopeptidase; AMC is released upon enzymatic cleavage by prolyl endopeptidase and its fluorescence can be used to quantify prolyl endopeptidase activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-294-4036 Supplier Cayman Chemical Supplier No. 4232010 MG
A fluorogenic substrate for prolyl endopeptidase; AMC is released upon enzymatic cleavage by prolyl endopeptidase and its fluorescence can be used to quantify prolyl endopeptidase activity; ex/em = 340-360/440-460 nm, respectively