Learn More
Medchemexpress LLC Cbz-L-Trp-OH | 7432-21-5 | 99.9% | 338.36 | 50 G
SDP
Catalog No. 5000453581
Encompass_Preferred
Cbz-L-Trp-OH is a competitive inhibitor of SCD (Stearoyl-CoA Desaturase) with an IC50 of 2.5 μM and a Ki of 2.1 μM. It is intended for research use only and is not sold to patients. The product appears as a solid, white to off-white in color.
  • Competitive inhibitor of SCD.
  • High purity: 99.93% (ee.: 100%).
  • Available in various quantities.
  • Provided with Data Sheet, COA, SDS, and Handling Instructions.
Catalog No. 50-004-53581 Supplier Medchemexpress LLC Supplier No. HYW01119750G
Cbz-L-Trp-OH is a competitive inhibitor of SCD (Stearoyl-CoA Desaturase) with an IC50 of 2.5 μM and a Ki of 2.1 μM. It is intended for research use only and is not sold to patients. The product appears as a solid, white to off-white in color.
  • Competitive inhibitor of SCD.
  • High purity: 99.93% (ee.: 100%).
  • Available in various quantities.
  • Provided with Data Sheet, COA, SDS, and Handling Instructions.