Learn More
Medchemexpress LLC Cbz-leucinyl-leucinyl-norleucinal | 133407-83-7 | 97.6% | 10 MG
SDP
Catalog No. 5000234307
Encompass_Preferred
Z-LLNle-CHO (Cbz-leucinyl-leucinyl-norleucinal) is a peptidic aldehyde inhibitor of γ-secretase used in biochemical and cell-based research. It is supplied as a high-purity solid suitable for in vitro assays.
  • Peptidic aldehyde inhibitor of γ-secretase.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight 475.62 g/mol and formula C26H41N3O5.
  • Supplied as a small, easily weighed solid for laboratory use.
  • Used to study apoptosis, proteasome, and protease pathways.
Catalog No. 50-002-34307 Supplier Medchemexpress LLC Supplier No. HY12023410MG
Z-LLNle-CHO (Cbz-leucinyl-leucinyl-norleucinal) is a peptidic aldehyde inhibitor of γ-secretase used in biochemical and cell-based research. It is supplied as a high-purity solid suitable for in vitro assays.
  • Peptidic aldehyde inhibitor of γ-secretase.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight 475.62 g/mol and formula C26H41N3O5.
  • Supplied as a small, easily weighed solid for laboratory use.
  • Used to study apoptosis, proteasome, and protease pathways.