Learn More
Medchemexpress LLC Cbz-Lys(Boc)-OH | 2389-60-8 | 99.4% | 380.44 | 10 G
SDP
Catalog No. 5000366893
Encompass_Preferred
Cbz-Lys(Boc)-OH is a protected amino acid primarily utilized in peptide synthesis, protein engineering, and drug development, particularly in research involving lysine residues.
  • Used in peptide synthesis.
  • Applicable in protein engineering.
  • Valuable for drug development.
  • Beneficial for research involving lysine residues.
Catalog No. 50-003-66893 Supplier Medchemexpress LLC Supplier No. HYW00903310G
Cbz-Lys(Boc)-OH is a protected amino acid primarily utilized in peptide synthesis, protein engineering, and drug development, particularly in research involving lysine residues.
  • Used in peptide synthesis.
  • Applicable in protein engineering.
  • Valuable for drug development.
  • Beneficial for research involving lysine residues.