Learn More
Medchemexpress LLC Cbz-Lys(Boc)-OH | 2389-60-8 | MFCD00062271 | 99.4% | 380.44 | 100 G
SDP
Catalog No. 5000453848
Encompass_Preferred
Cbz-Lys(Boc)-OH is a protected amino acid primarily used in peptide synthesis, protein engineering, and drug development, particularly in research involving lysine residues.
  • Utilized in peptide synthesis
  • Utilized in protein engineering
  • Utilized in drug development, especially in research focused on lysine residues
Catalog No. 50-004-53848 Supplier Medchemexpress LLC Supplier No. HYW009033100G
Cbz-Lys(Boc)-OH is a protected amino acid primarily used in peptide synthesis, protein engineering, and drug development, particularly in research involving lysine residues.
  • Utilized in peptide synthesis
  • Utilized in protein engineering
  • Utilized in drug development, especially in research focused on lysine residues