Learn More
Medchemexpress LLC Cbz-Lys(Boc)-OH | 2389-60-8 | MFCD00062271 | 99.4% | 380.44 | 50 G
SDP
Catalog No. 5000453846
Encompass_Preferred
Cbz-Lys(Boc)-OH is a protected amino acid primarily utilized in peptide synthesis, protein engineering, and drug development, particularly in research involving lysine residues.
  • Used in peptide synthesis, protein engineering, and drug development.
  • Especially useful in research involving lysine residues.
  • Solid at less than 64 °C, liquid at greater than 67 °C.
  • Off-white to light yellow color.
  • Ships at room temperature (continental US).
  • Stable for 3 years at room temperature; in solvent, stable for 2 years at -80 °C or 1 year at -20 °C.
Catalog No. 50-004-53846 Supplier Medchemexpress LLC Supplier No. HYW00903350G
Cbz-Lys(Boc)-OH is a protected amino acid primarily utilized in peptide synthesis, protein engineering, and drug development, particularly in research involving lysine residues.
  • Used in peptide synthesis, protein engineering, and drug development.
  • Especially useful in research involving lysine residues.
  • Solid at less than 64 °C, liquid at greater than 67 °C.
  • Off-white to light yellow color.
  • Ships at room temperature (continental US).
  • Stable for 3 years at room temperature; in solvent, stable for 2 years at -80 °C or 1 year at -20 °C.