Learn More
Medchemexpress LLC Cbz-NH-PEG12-C2-acid | 1334177-88-6 | 95.7% | C35H61NO16 | 10MG
SDP
Catalog No. 5000259056
Encompass_Preferred
Cbz-NH-PEG12-C2-acid is a PEG-based PROTAC linker (C35H61NO16, MW 751.86 g·mol⁻¹) that serves as a hydrophilic spacer between ligand moieties. It bears a Cbz-protected amine and a terminal carboxylic acid, providing orthogonal functional groups for coupling and downstream modifications in medicinal chemistry and PROTAC assembly.
  • PEG12 spacer provides hydrophilicity and conformational flexibility.
  • Cbz-protected amine enables selective deprotection and subsequent functionalization.
  • Terminal carboxylic acid is compatible with amide and ester coupling chemistries.
  • Optimized for use as a linker in PROTAC synthesis and related conjugation strategies.
  • Supplied with high purity suitable for synthetic applications.
Catalog No. 50-002-59056 Supplier Medchemexpress LLC Supplier No. HY13335810MG
Cbz-NH-PEG12-C2-acid is a PEG-based PROTAC linker (C35H61NO16, MW 751.86 g·mol⁻¹) that serves as a hydrophilic spacer between ligand moieties. It bears a Cbz-protected amine and a terminal carboxylic acid, providing orthogonal functional groups for coupling and downstream modifications in medicinal chemistry and PROTAC assembly.
  • PEG12 spacer provides hydrophilicity and conformational flexibility.
  • Cbz-protected amine enables selective deprotection and subsequent functionalization.
  • Terminal carboxylic acid is compatible with amide and ester coupling chemistries.
  • Optimized for use as a linker in PROTAC synthesis and related conjugation strategies.
  • Supplied with high purity suitable for synthetic applications.