Learn More
Medchemexpress LLC Cbz-NH-PEG4-C2-acid | 756526-00-8 | MFCD11041139 | 95.0% | 399.44 g/mol | C19H29NO8 | 250 MG
SDP
Catalog No. 5000302460
Encompass_Preferred
Cbz-NH-PEG4-C2-acid is a PEG-based linker designed for PROTAC synthesis. It features a Cbz-protected amino group and a terminal carboxylic acid separated by a four-unit ethylene glycol spacer, providing solubility and flexibility for conjugation. The compound is intended for use in medicinal chemistry to connect ligands and enable targeted protein degrader construction.
  • Cbz-protected amino group for selective deprotection
  • Terminal carboxylic acid enables standard amide-coupling chemistries
  • PEG4 spacer imparts flexibility and improves solubility
  • Suitable for PROTAC linker construction and linker optimization studies
  • Available in common laboratory pack sizes for synthesis workflows
Catalog No. 50-003-02460 Supplier Medchemexpress LLC Supplier No. 250MGHYW019796
Cbz-NH-PEG4-C2-acid is a PEG-based linker designed for PROTAC synthesis. It features a Cbz-protected amino group and a terminal carboxylic acid separated by a four-unit ethylene glycol spacer, providing solubility and flexibility for conjugation. The compound is intended for use in medicinal chemistry to connect ligands and enable targeted protein degrader construction.
  • Cbz-protected amino group for selective deprotection
  • Terminal carboxylic acid enables standard amide-coupling chemistries
  • PEG4 spacer imparts flexibility and improves solubility
  • Suitable for PROTAC linker construction and linker optimization studies
  • Available in common laboratory pack sizes for synthesis workflows