Learn More
Medchemexpress LLC Cbz-NH-PEG6-C2-acid | 1334177-80-8 | 100 MG
SDP
Catalog No. 5000339955
Encompass_Preferred
Cbz-NH-PEG6-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two different ligands: one for an E3 ubiquitin ligase and another for the target protein. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Designed to selectively degrade target proteins
Catalog No. 50-003-39955 Supplier Medchemexpress LLC Supplier No. HY130443100MG
Cbz-NH-PEG6-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two different ligands: one for an E3 ubiquitin ligase and another for the target protein. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Designed to selectively degrade target proteins