Learn More
Medchemexpress LLC CC-115 hydrochloride | 1300118-55-1 | 98.0% | 372.81 | 100 MG
SDP
Catalog No. 5000393467
Encompass_Preferred
CC-115 hydrochloride is a potent dual DNA-PK and mTOR kinase inhibitor that blocks both mTORC1 and mTORC2 signaling. It inhibits PC-3 cell proliferation and demonstrates selectivity against other protein kinases and PIKKs, including good selectivity against PI3K-alpha, ATR, and ATM. It shows good in vivo PK profiles across multiple species.
  • Potent and dual DNA-PK and mTOR kinase inhibitor (IC50s of 13 nM and 21 nM respectively)
  • Blocks both mTORC1 and mTORC2 signaling
  • Inhibits PC-3 cells proliferation with an IC50 of 138 nM
  • Demonstrates selectivity against other protein kinases and PIKKs
  • Shows good in vivo PK profiles across multiple species (53% in mouse, 76% in rat, and ~100% in dog oral bioavailability)
  • Leads to significant tumor volume reductions and sustains inhibition in vivo
Catalog No. 50-003-93467 Supplier Medchemexpress LLC Supplier No. HY16962A100MG
CC-115 hydrochloride is a potent dual DNA-PK and mTOR kinase inhibitor that blocks both mTORC1 and mTORC2 signaling. It inhibits PC-3 cell proliferation and demonstrates selectivity against other protein kinases and PIKKs, including good selectivity against PI3K-alpha, ATR, and ATM. It shows good in vivo PK profiles across multiple species.
  • Potent and dual DNA-PK and mTOR kinase inhibitor (IC50s of 13 nM and 21 nM respectively)
  • Blocks both mTORC1 and mTORC2 signaling
  • Inhibits PC-3 cells proliferation with an IC50 of 138 nM
  • Demonstrates selectivity against other protein kinases and PIKKs
  • Shows good in vivo PK profiles across multiple species (53% in mouse, 76% in rat, and ~100% in dog oral bioavailability)
  • Leads to significant tumor volume reductions and sustains inhibition in vivo