Learn More
Medchemexpress LLC CC-115 (hydrochloride) | 1300118-55-1 | 98.0% | 372.81 | 25 MG
SDP
Catalog No. 5000394913
Encompass_Preferred
CC-115 hydrochloride is a potent dual inhibitor of DNA-PK and mTOR kinase. It effectively blocks both mTORC1 and mTORC2 signaling pathways, with IC50 values of 13 nM and 21 nM respectively. This product is intended for research use only.
  • Potent and dual DNA-PK and mTOR kinase inhibitor
  • Blocks both mTORC1 and mTORC2 signaling
  • Exhibits good in vivo PK profiles across multiple species
  • Demonstrates significant tumor volume reductions in tested models
  • Sustains inhibition for prolonged periods
  • Available in various sizes, including solid and pre-made solutions
Catalog No. 50-003-94913 Supplier Medchemexpress LLC Supplier No. 25MGHY16962A
CC-115 hydrochloride is a potent dual inhibitor of DNA-PK and mTOR kinase. It effectively blocks both mTORC1 and mTORC2 signaling pathways, with IC50 values of 13 nM and 21 nM respectively. This product is intended for research use only.
  • Potent and dual DNA-PK and mTOR kinase inhibitor
  • Blocks both mTORC1 and mTORC2 signaling
  • Exhibits good in vivo PK profiles across multiple species
  • Demonstrates significant tumor volume reductions in tested models
  • Sustains inhibition for prolonged periods
  • Available in various sizes, including solid and pre-made solutions