Learn More
Medchemexpress LLC CC-115 hydrochloride | 1300118-55-1 | 98.01% | 372.81 | 1 ML
SDP
Catalog No. 5000393794
Encompass_Preferred
CC-115 hydrochloride is a potent, dual DNA-PK and mTOR kinase inhibitor. It targets both mTORC1 and mTORC2 signaling, making it a valuable tool for research applications. This compound has demonstrated significant inhibitory effects against PC-3 cell proliferation and exhibits good in vivo pharmacokinetic profiles, including high oral bioavailability in various animal models.
  • Potent DNA-PK inhibitor (IC50 of 13 nM)
  • Potent mTOR kinase inhibitor (IC50 of 21 nM)
  • Blocks both mTORC1 and mTORC2 signaling
  • Inhibits PC-3 cell proliferation with an IC50 of 138 nM
  • Exhibits good in vivo pharmacokinetic profiles
  • Demonstrates high oral bioavailability in mouse, rat, and dog
  • Sustains inhibition through 24 hours
  • Shows tumor volume reductions in vivo studies
Catalog No. 50-003-93794 Supplier Medchemexpress LLC Supplier No. HY16962A10MM/1ML
CC-115 hydrochloride is a potent, dual DNA-PK and mTOR kinase inhibitor. It targets both mTORC1 and mTORC2 signaling, making it a valuable tool for research applications. This compound has demonstrated significant inhibitory effects against PC-3 cell proliferation and exhibits good in vivo pharmacokinetic profiles, including high oral bioavailability in various animal models.
  • Potent DNA-PK inhibitor (IC50 of 13 nM)
  • Potent mTOR kinase inhibitor (IC50 of 21 nM)
  • Blocks both mTORC1 and mTORC2 signaling
  • Inhibits PC-3 cell proliferation with an IC50 of 138 nM
  • Exhibits good in vivo pharmacokinetic profiles
  • Demonstrates high oral bioavailability in mouse, rat, and dog
  • Sustains inhibition through 24 hours
  • Shows tumor volume reductions in vivo studies