Learn More
Medchemexpress LLC HY-16962A 5mg Medchemexpress, CC-115 (hydrochloride) CAS:1300118-55-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16962A5MG
Encompass_Preferred
Medchemexpress, HY-16962A 5mg CC-115 (hydrochloride) CAS:1300118-55-1 CC-115 hydrochloride is a potent and dual DNA-PK and mTOR kinase inhibitor with IC50s of 13 nM and 21 nM, respectively. CC-115 blocks both mTORC1 and mTORC2 signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1919