Learn More
Huber USA CC-118A, Clear View Circulator Bath
Supplier:  Huber USA 2053.0002.01
Encompass_Preferred
‐ Temperature Range: (15) 25°C ... +100°C‐ Heating capacity: 1,5 kW‐ Pressure pump max.: 27 l/min / 0,7 bar‐ Suction pump max.: 22 l/min / 0,4 bar‐ Bath volume: 18 l‐ Power supply: 110V 1~ 50/60Hz
Catalog No. 50-110-8632