Learn More
CELLTREAT Scientific Products Permeable Cell Culture Inserts, Packed in 24 Well Plate, Hanging, PC, 8.0m, Sterile, 24/CS
SDP
Catalog No. 501535161
Encompass_Preferred
CELLTREAT Scientific Products Permeable Cell Culture Inserts. Permeable Cell Culture Inserts mimic in vivo conditions. Membrane remains suspended above well bottom, allowing co-culture: grow one cell line inside insert, and another below in the well. Ideal for drug transfer assays, transport studies, migration assays, and cell-cell interaction studies. Inserts center in wells, preventing membrane from touch-ing sides and wicking between insert and side wall. 0.4um pore size is ideal for co-culture and drug transport studies. 3.0 and 8.0um pore size is ideal for angiogenesis and chemotaxis experiments. Non -Treated Plates sold separately; carrier plate is Tissue Culture Treated. Plate manufactured from high clarity virgin polysytrene with polycarbonate membrane insert. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile with 12 Inserts/plate, 2 Plates/case, 24 Inserts/case.
Catalog No. 50-153-5161 Supplier CELLTREAT Scientific Products Supplier No. 230633
CELLTREAT Scientific Products Permeable Cell Culture Inserts. Permeable Cell Culture Inserts mimic in vivo conditions. Membrane remains suspended above well bottom, allowing co-culture: grow one cell line inside insert, and another below in the well. Ideal for drug transfer assays, transport studies, migration assays, and cell-cell interaction studies. Inserts center in wells, preventing membrane from touch-ing sides and wicking between insert and side wall. 0.4um pore size is ideal for co-culture and drug transport studies. 3.0 and 8.0um pore size is ideal for angiogenesis and chemotaxis experiments. Non -Treated Plates sold separately; carrier plate is Tissue Culture Treated. Plate manufactured from high clarity virgin polysytrene with polycarbonate membrane insert. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile with 12 Inserts/plate, 2 Plates/case, 24 Inserts/case.