Learn More
Medchemexpress LLC CC-3240 | 2922675-91-8 | 98.2% | 917.17 | 5 MG
SDP
Catalog No. 5000392109
Encompass_Preferred
CC-3240 is a molecular glue degrader of CaMKK2, based on compound 13, with an IC50 of 9 nM. It is intended for research use only and is not for patient treatment.
  • Antiproliferative activity against human A-427 cells.
  • Reduction in cell growth after 48 hours incubation by CellTiter assay (EC50 > 8 μM).
Catalog No. 50-003-92109 Supplier Medchemexpress LLC Supplier No. HY1630645MG
CC-3240 is a molecular glue degrader of CaMKK2, based on compound 13, with an IC50 of 9 nM. It is intended for research use only and is not for patient treatment.
  • Antiproliferative activity against human A-427 cells.
  • Reduction in cell growth after 48 hours incubation by CellTiter assay (EC50 > 8 μM).