Learn More
Medchemexpress LLC CC-3240 | 2922675-91-8 | 98.23% | 917.17 | 1 MG
SDP
Catalog No. 5000392110
Encompass_Preferred
CC-3240 (compound 13) is a molecular glue degrader of CaMKK2 based on CC-8977, with an IC50 of 9 nM. It is for research use only and not sold to patients.
  • Molecular glue degrader of CaMKK2
  • IC50 of 9 nM
  • Antiproliferative activity against human A-427 cells (EC50 > 8 μM)
Catalog No. 50-003-92110 Supplier Medchemexpress LLC Supplier No. HY1630641MG
Add to Cart
Add to Cart
CC-3240 (compound 13) is a molecular glue degrader of CaMKK2 based on CC-8977, with an IC50 of 9 nM. It is for research use only and not sold to patients.
  • Molecular glue degrader of CaMKK2
  • IC50 of 9 nM
  • Antiproliferative activity against human A-427 cells (EC50 > 8 μM)