Learn More
Medchemexpress LLC CC-401 hydrochloride | 1438391-30-0 | 99.35% | 25 MG
SDP
Catalog No. 5000342368
Encompass_Preferred
CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM. It exhibits at least 40-fold selectivity for JNK compared with other related kinases, including p38, extracellular signal-regulated kinase (ERK), inhibitor of κB kinase (IKK2), protein kinase C, Lck, and zeta-associated protein of 70 kDa (ZAP70).
  • Competitively binds the ATP binding site in JNK.
  • Inhibits phosphorylation of the N-terminal activation domain of the transcription factor c-Jun.
  • Demonstrates specificity in vitro using osmotic stress of the HK-2 human tubular epithelial cell line.
  • Inhibits sorbitol-induced phosphorylation of c-Jun in a dosage-dependent manner.
  • Does not prevent sorbitol-induced phosphorylation of JNK, p38, or ERK induced by sorbitol.
  • Provides specific JNK inhibition in cell-based assays at 1 to 5 μM.
Catalog No. 50-003-42368 Supplier Medchemexpress LLC Supplier No. 25MGHY13022
CC-401 hydrochloride is a potent inhibitor of all three forms of JNK with Ki of 25 to 50 nM. It exhibits at least 40-fold selectivity for JNK compared with other related kinases, including p38, extracellular signal-regulated kinase (ERK), inhibitor of κB kinase (IKK2), protein kinase C, Lck, and zeta-associated protein of 70 kDa (ZAP70).
  • Competitively binds the ATP binding site in JNK.
  • Inhibits phosphorylation of the N-terminal activation domain of the transcription factor c-Jun.
  • Demonstrates specificity in vitro using osmotic stress of the HK-2 human tubular epithelial cell line.
  • Inhibits sorbitol-induced phosphorylation of c-Jun in a dosage-dependent manner.
  • Does not prevent sorbitol-induced phosphorylation of JNK, p38, or ERK induced by sorbitol.
  • Provides specific JNK inhibition in cell-based assays at 1 to 5 μM.