Learn More
Selleck Chemical LLC CC-401 Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S67305MG
Encompass_Preferred
CC-401 is a potent inhibitor of JNK with at least 40-fold selectivity against other related kinases
Catalog No. 50-000-02022