Learn More
Selleck Chemical LLC CC-401 Hydrochloride
SDP
Catalog No. 5000002904
Encompass_Preferred
CC-401 is a potent inhibitor of JNK with at least 40-fold selectivity against other related kinases
Catalog No. 50-000-02904 Supplier Selleck Chemical LLC Supplier No. S67302MG
CC-401 is a potent inhibitor of JNK with at least 40-fold selectivity against other related kinases