Learn More
Medchemexpress LLC HY-108709 5mg Medchemexpress, CC-671 CAS:1618658-88-0 Purity:>98%
SDP
Catalog No. 501968618
Encompass_Preferred
Medchemexpress, HY-108709 5mg CC-671 CAS:1618658-88-0 CC-671 is a dual TTK protein kinase/CDC2-like kinase (CLK2) inhibitor with IC50s of 0.005 and 0.006 μM for TTK and CLK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8618 Supplier Medchemexpress LLC Supplier No. HY1087095MG
Medchemexpress, HY-108709 5mg CC-671 CAS:1618658-88-0 CC-671 is a dual TTK protein kinase/CDC2-like kinase (CLK2) inhibitor with IC50s of 0.005 and 0.006 μM for TTK and CLK2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.