Learn More
Medchemexpress LLC HY-101488 10mg Medchemexpress, CC-885 CAS:1010100-07-8 Purity:>98%
SDP
Catalog No. 502028660
Encompass_Preferred
Medchemexpress, HY-101488 10mg CC-885 CAS:1010100-07-8 CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8660 Supplier Medchemexpress LLC Supplier No. HY10148810MG
Medchemexpress, HY-101488 10mg CC-885 CAS:1010100-07-8 CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.