Learn More
Selleck Chemical LLC CC-885-5mg
SDP
Catalog No. 502389792
Encompass_Preferred
CC-885 is a novel cereblon (CRBN) modulator. CC-885 selectively promotes CRBN- and p97-dependent PLK1 ubiquitination and degradation.
Catalog No. 50-238-9792 Supplier Selleck Chemical LLC Supplier No. S83005MG
CC-885 is a novel cereblon (CRBN) modulator. CC-885 selectively promotes CRBN- and p97-dependent PLK1 ubiquitination and degradation.