Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cc-885-ch2-peg1-nh-ch3 | 2722698-03-3 | 99.2% | 528.00 | C26H30ClN5O5 | 25 MG
SDP

Catalog No. 5000203774
Encompass_Preferred

CC-885-CH2-PEG1-NH-CH3 is a small-molecule neoDegrader used as the cytotoxic payload in antibody-based neoDegrader conjugates (AnDCs) to enable targeted protein degradation via the E3 ubiquitin ligase pathway. It is supplied for laboratory research use.

  • Used in synthesis of antibody neoDegrader conjugates.
  • Enables targeted protein degradation via the E3 ubiquitin ligase pathway.
  • Molecular formula C26H30ClN5O5; molecular weight 528.00 g/mol.
  • Purity 99.2% and solid, white to off-white appearance.
  • Storage: 4°C (sealed); in solvent, -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-03774 Supplier Medchemexpress LLC Supplier No. HY14544925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CC-885-CH2-PEG1-NH-CH3 is a small-molecule neoDegrader used as the cytotoxic payload in antibody-based neoDegrader conjugates (AnDCs) to enable targeted protein degradation via the E3 ubiquitin ligase pathway. It is supplied for laboratory research use.

  • Used in synthesis of antibody neoDegrader conjugates.
  • Enables targeted protein degradation via the E3 ubiquitin ligase pathway.
  • Molecular formula C26H30ClN5O5; molecular weight 528.00 g/mol.
  • Purity 99.2% and solid, white to off-white appearance.
  • Storage: 4°C (sealed); in solvent, -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.