Learn More
Medchemexpress LLC HY-132304 5mg , CC-90005 CAS:1799574-70-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1323045MG
Encompass_Preferred
HY-132304 5mg CC-90005 CAS: 1799574-70-1 CC-90005 is a potent, selective and orally active inhibitor of protein kinase C-θ (PKC-θ), with an IC50 of 8 nM. CC-90005 shows selectivity for PKC-θ over PKC-δ (IC50=4440 nM). CC-90005 can inhibit T cell activation by IL-2 expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1372