Learn More
Medchemexpress LLC HY-129395 5mg Medchemexpress, CC-92480 CAS:2259648-80-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1293955MG
Encompass_Preferred
Medchemexpress, HY-129395 5mg CC-92480 CAS:2259648-80-9 CC-92480 is a cereblon E3 ubiquitin ligase modulating drug (CELMoD). CC-92480 shows high affinity to cereblon, resulting in potent antimyeloma activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8931