Learn More
Waters Corp Waters Heat Exchanger Tubing, Countercurrent, Post-Column Reaction Module
Catalog No. NC2868616 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This countercurrent heat exchanger tubing is a key component of the Post-Column Reaction Module (PCRM). It is designed to efficiently manage temperature within the module, ensuring optimal conditions for chemical reactions and detector performance.
  • Pre-warms post-column reagents and HPLC column effluent.
  • Transfers heat from reactor effluent to incoming streams for uniform reactions.
  • Cools reactor effluent to nearly ambient temperature.
  • Improves detector performance.
  • Protects reagent solutions and unstable derivatives from thermal degradation.
  • Minimizes background fluorescence.
Catalog No. NC2868616 Supplier Waters Corp Supplier No. 700001051
This countercurrent heat exchanger tubing is a key component of the Post-Column Reaction Module (PCRM). It is designed to efficiently manage temperature within the module, ensuring optimal conditions for chemical reactions and detector performance.
  • Pre-warms post-column reagents and HPLC column effluent.
  • Transfers heat from reactor effluent to incoming streams for uniform reactions.
  • Cools reactor effluent to nearly ambient temperature.
  • Improves detector performance.
  • Protects reagent solutions and unstable derivatives from thermal degradation.
  • Minimizes background fluorescence.