Learn More
Huber USA CC-K25, Refrigerated Heating Bath
Catalog No. 501108676
Encompass_Preferred
‐ Temperature range: -30°C ... +200°C - Cooling capacity (20°C): 0,4 kW‐ Cooling capacity (0°C): 0,35 kW‐ Heating capacity: 1,5 kW‐ Pressure pump max.: 27 l/min / 0,7 bar‐ Suction pump max. : 22 l/min / 0,4 bar‐ Bath volume: 25 l‐ Power supply: 115V 1~ 60Hz
Catalog No. 50-110-8676 Supplier Huber USA Supplier No. 2012.0024.01
Add to Cart
Add to Cart
Request a Quote
‐ Temperature range: -30°C ... +200°C - Cooling capacity (20°C): 0,4 kW‐ Cooling capacity (0°C): 0,35 kW‐ Heating capacity: 1,5 kW‐ Pressure pump max.: 27 l/min / 0,7 bar‐ Suction pump max. : 22 l/min / 0,4 bar‐ Bath volume: 25 l‐ Power supply: 115V 1~ 60Hz