Learn More
Sigma-Aldrich CC/Mount™
Catalog No. 01258868 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
30 mL
Catalog No. Volume (Metric)
01-258-868 30 mL
1 options
Catalog No. 01-258-868 Supplier Sigma-Aldrich Supplier No. C936830ML
Add to Cart
Add to Cart
Aqueous-based permanent mounting medium.
CC/Mount is an aqueous based permanent mounting medium, similar to Crystal Mount™, for the permanent preservation of tissue sections stained with peroxidase and alkaline phosphatase based systems as well as with various fluorescent dyes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 30 mL