Learn More
Diagnostic Biosystems CC/MOUNT CC/Mount™
Supplier:  Diagnostic Biosystems K002
Encompass_Preferred
CC/Mount is an aqueous mounting medium with very high refractive index. When applied to the stained tissue sections, specimens can be permanently mounted without chromogens fading. Due to the superior refractive index of CC/Mount, tissues mounted in this medium look like dehydrated specimens. No coverslipping is required with CC/Mount. However, if coverslipping is desired, dried CC/Mount can be post-mounted by using an organic based mounting medium. Advantages of CC/Mount are numerous: no coverslip, no exposure to the organic fumes, permanent storage of slides and high resolution of tissue specimens. CC/Mount is compatible with chromogens lik AEC, DAB, Fast Red, BCIP/NBT, BCIP/INT and fluorescent dyes like FITC and phycobiliproteins. The high pH of CC/Mount ensures increased stability of fluorescence.\n\nThis product is designed for permanent mounting of tissue specimens stained with peroxidase and alkaline phosphatase-based systems and various fluorescent dyes.
Catalog No. NC2047659