Learn More
Chondrex Inc CC PORCINE COL1, SOL, 50 MG
Catalog No. 501526665
Encompass_Preferred
>95% porcine type I tropocollagen dissolved in 0.01M HCl. Solution form: 4 mg/mL x 12.5 mL. Sterile Filtered. Endotoxin level: < 1EU/ml. Intended for use in making 3D cell culture scaffolds. Contact Chondrex, Inc. at support@chondrex.com for technical assistance in making 3D cell culture scaffolds.
Catalog No. 50-152-6665 Supplier Chondrex Inc Supplier No. 1203
Add to Cart
Add to Cart
>95% porcine type I tropocollagen dissolved in 0.01M HCl. Solution form: 4 mg/mL x 12.5 mL. Sterile Filtered. Endotoxin level: < 1EU/ml. Intended for use in making 3D cell culture scaffolds. Contact Chondrex, Inc. at support@chondrex.com for technical assistance in making 3D cell culture scaffolds.