Learn More
Beckman Coulter Multisizer 4e
Catalog No. NC2291480
Encompass_Preferred
The Beckman Coulter Multisizer 4e is an automated cell counter and analyzer that uses Coulter Principle technology to provide accurate size distribution and concentration measurements of cells and particles from 0.4 to 1,200 microns in diameter. It offers a dynamic range up to 1x10^9 particles/mL and can measure and analyze multiple cell types and samples in one run. The Multisizer 4e streamlines particle characterization workflows and ensures consistent, reproducible results. Its modular design allows customization of the system to meet specific application needs.
This item is not returnable. View return policy
Catalog No. NC2291480 Supplier Beckman Coulter Supplier No. C72517
The Beckman Coulter Multisizer 4e is an automated cell counter and analyzer that uses Coulter Principle technology to provide accurate size distribution and concentration measurements of cells and particles from 0.4 to 1,200 microns in diameter. It offers a dynamic range up to 1x10^9 particles/mL and can measure and analyze multiple cell types and samples in one run. The Multisizer 4e streamlines particle characterization workflows and ensures consistent, reproducible results. Its modular design allows customization of the system to meet specific application needs.