Learn More
Beckman Coulter Multisizer 4e
Supplier:  Beckman Coulter C72518
Encompass_Preferred
The Beckman Coulter Multisizer 4e is a cell counter and analyzer that uses Coulter Principle technology to provide accurate size distribution and concentration measurements of cells and particles from 0.4 to 1,200 microns in diameter. It offers a dynamic range up to 1x10^9 particles/mL and can measure and analyze up to 32 samples simultaneously with high precision and reproducibility. The modular design allows customization for specific applications. Data analysis software is included for displaying size distributions and analyzing results. The system is designed for use in biopharmaceutical, chemical, food, beverage and other industries requiring particle characterization.
Catalog No. NC2291481