Learn More
Beckman Coulter Access 2 Immunoassay System Operator's Guide
Supplier:  Beckman Coulter C72515
Encompass_Preferred
The Beckman Coulter AU480 Chemistry Analyzer is an automated clinical chemistry analyzer designed for high-volume laboratories. It utilizes ion selective electrode and photometric technologies to provide test results for routine chemistries, specific proteins, therapeutic drugs, and more. The AU480 has a throughput of up to 1200 tests per hour and features a sample loading capacity of 150 samples. It requires minimal maintenance while providing quick and accurate test results. The AU480 analyzer streamlines workflow and increases productivity in busy laboratories.
Catalog No. NC2291479