Learn More
Bullard™ CC20 Breathing Tube Assemblies for Hoods
SDP
Catalog No. 19065672 Shop All Bullard Company Products
Change view
Click to view available options
Connection Type:
0.25 in. Industrial Interchange
0.25 in. Snap-tite
0.25 in. Snap-tite Brass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
19-065-672 0.25 in. Snap-tite Brass
17-987-503 0.25 in. Industrial Interchange
19-021-319 0.25 in. Snap-tite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-065-672 Supplier Bullard™ Supplier No. V33
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with CC20 Series respirators.
  • Component for use with CC20 air-supplied systems
  • 0.25 in. connection in snap-tite or industrial interchange versions
  • Quick-disconnect
Specifications
Connection Type 0.25 in. Snap-tite Brass
Material Brass
Product Type Breathing Tube
For Use With (Equipment) CC20 Series Airline Compressed Air Respirator
Includes Quick-disconnect nipple
Model V33
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.