Learn More
ABclonal Technology CCAR1 Rabbit pAb
SDP
Catalog No. 5000097801
Encompass_Preferred
Enables RNA polymerase II cis-regulatory region sequence-specific DNA binding activity, nuclear receptor coactivator activity, and transcription corepressor activity. Involved in positive regulation of cell migration and positive regulation of cell population proliferation. Acts upstream of or within positive regulation of apoptotic process. Located in nuclear envelope lumen.
Catalog No. 50-000-97801 Supplier ABclonal Technology Supplier No. A1359520UL
Enables RNA polymerase II cis-regulatory region sequence-specific DNA binding activity, nuclear receptor coactivator activity, and transcription corepressor activity. Involved in positive regulation of cell migration and positive regulation of cell population proliferation. Acts upstream of or within positive regulation of apoptotic process. Located in nuclear envelope lumen.