Learn More
Aviva Systems Biology Corp CCBE1 Antibody - middle region (ARP52564_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52564_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCBE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCBE1. Alternate names: FLJ30681, MGC50861ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62894