Learn More
Aviva Systems Biology Corp CCBP2 Antibody - N-terminal region (ARP30776_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30776_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCBP2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CCBP2. Alternate names: CCR10, CCR9, CMKBR9, D6, MGC126678, MGC138250, hD6, CCBP2ApplicationIHC, WB. Species reactivity: Human, Rabbit
Catalog No. 50-014-54166