Learn More
Electron Microscopy Sciences EMS CCC Carbon Adhesive 30 GR
SDP
Catalog No. 50980324
Encompass_Preferred
An electrically Carbon Conductive Cement for specimen mounting in all SEM work. After drying of the cement, immediate investigation of conductive specimens is possible. Non-conductive specimens need only to be coated with carbon or metal. Thinner is available if the cement viscosity is too thick.
Catalog No. 50-980-324 Supplier Electron Microscopy Sciences Supplier No. 12664
An electrically Carbon Conductive Cement for specimen mounting in all SEM work. After drying of the cement, immediate investigation of conductive specimens is possible. Non-conductive specimens need only to be coated with carbon or metal. Thinner is available if the cement viscosity is too thick.