Learn More
Electron Microscopy Sciences EMS CCC Thinner 30 ML
SDP
Catalog No. 50980325
Encompass_Preferred
An electrically Carbon Conductive Cement for specimen mounting in all SEM work. After drying of the cement, immediate investigation of conductive specimens is possible. Non-conductive specimens need only to be coated with carbon or metal. Thinner is available if the cement viscosity is too thick.
Catalog No. 50-980-325 Supplier Electron Microscopy Sciences Supplier No. 12665
An electrically Carbon Conductive Cement for specimen mounting in all SEM work. After drying of the cement, immediate investigation of conductive specimens is possible. Non-conductive specimens need only to be coated with carbon or metal. Thinner is available if the cement viscosity is too thick.