Learn More
ATCC CCD 841 CoN; Colon; Human (Homo sapiens)
Supplier:  ATCC CRL1790
Encompass
CCD 841 CoN cells were isolated from normal human colon tissue. The cells resemble epithelial cells; however, the cells do not contain keratin and definitive evidence of epithelial origin is lacking.
Catalog No. 50-238-4038
 Restricted Item