Learn More
ATCC CCD-1079Sk; Foreskin; Human (Homo Sapiens)
Catalog No. 502383246
Encompass
CCD-1079Sk are fibroblasts that were isolated from the skin of a normal male neonate donor. It can be used in cancer research or to derive induced pluripotent stem cells.
Catalog No. 50-238-3246 Supplier ATCC Supplier No. CRL2097
CCD-1079Sk are fibroblasts that were isolated from the skin of a normal male neonate donor. It can be used in cancer research or to derive induced pluripotent stem cells.