Learn More
ATCC CCD-16 Lu; Lung Fibroblast; Human (Homo sapiens)
Supplier:  ATCC CCL204
Encompass
CCD-16Lu fibroblast cell line is derived from normal lung tissue from a patient who died of a class 1 - 2 astrocytoma.
Catalog No. 50-238-5021