Learn More
ATCC CCD-19Lu; Lung Fibroblast; Human (Homo sapiens)
Catalog No. 502381837
Encompass
CCD-19Lu is a fibroblast cell line that was isolated from the lung of a normal White, 20-year-old, female patient. This cell line was deposited by S Dilworth.
Catalog No. 50-238-1837 Supplier ATCC Supplier No. CCL210
CCD-19Lu is a fibroblast cell line that was isolated from the lung of a normal White, 20-year-old, female patient. This cell line was deposited by S Dilworth.