Learn More
INTACT GENOMICS INC ig ccdB Resist™ Chemically Competent Cells 12x50μl
Catalog No. 5000082855
Encompass
Intact Genomics (ig) ccdB Resist™ Chemically Competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. ccdB Resist™ cells are capable of propagation of plasmids containing the ccdB gene. Intact Genomics ccdB Resist™ cells provide superb transformation efficiency, allowing for increased opportunity for experimental success.ig ccdB Resist™ Chemically Competent cells have the following features: Resistance to the ccdB gene product, as well as the T1 and T5 phage High transformation efficiency endA1 mutation increases plasmid yield significantly Capable of blue/white screening
Catalog No. 50-000-82855 Supplier INTACT GENOMICS INC Supplier No. 106912
Intact Genomics (ig) ccdB Resist™ Chemically Competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. ccdB Resist™ cells are capable of propagation of plasmids containing the ccdB gene. Intact Genomics ccdB Resist™ cells provide superb transformation efficiency, allowing for increased opportunity for experimental success.ig ccdB Resist™ Chemically Competent cells have the following features: Resistance to the ccdB gene product, as well as the T1 and T5 phage High transformation efficiency endA1 mutation increases plasmid yield significantly Capable of blue/white screening