Learn More
Aviva Systems Biology Corp CCDC102B Antibody - N-terminal region (ARP62777_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62777_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC102B. It was validated on Western Blot. . Alternate names: ACY1L, AN, C18orf14, DKFZp434K1426, DKFZp686I08254, FLJ23594, HsT1731, MGC161726, MGC161728ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-767