Learn More
Aviva Systems Biology Corp CCDC105 Antibody - C-terminal region (ARP53239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC105. It was validated on Western Blot. . Alternate names: FLJ40365ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-813