Learn More
Aviva Systems Biology Corp CFAP53 Antibody - N-terminal region (ARP55454_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55454_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC11. Alternate names: FLJ32743ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig
Catalog No. 50-014-61511