Learn More
Aviva Systems Biology Corp CCDC110 Antibody - middle region (ARP55544_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55544_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC110. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CCDC110. Alternate names: KM-HN-1, KMHN1, MGC33607, CT52ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-053